missing translation for 'onlineSavingsMsg'
Learn More

Cypermethrin, 96%

Catalog No. AC456310250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456310250 25 g
AC456310050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456310250 Supplier Thermo Scientific Chemicals Supplier No. 456310250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52315-07-8 | C22H19Cl2NO3 | 416.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52315-07-8
Molecular Formula C22H19Cl2NO3
Molecular Weight (g/mol) 416.3
InChI Key KAATUXNTWXVJKI-UHFFFAOYSA-N
Synonym cypermethrin, barricade, ripcord, ustaad, ammo, supercypermethrin, cymbush, alpha-cypermethrin, cypermethrine, agrothrin
PubChem CID 2912
ChEBI CHEBI:4042
IUPAC Name [cyano-(3-phenoxyphenyl)methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMILES CC1(C(C1C(=O)OC(C#N)C2=CC(=CC=C2)OC3=CC=CC=C3)C=C(Cl)Cl)C

Specifications

Density 1.1200g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Mixture of isomers) (HPLC)
Quantity 25 g
Specific Gravity 1.12
Solubility Information Solubility in water: insoluble.
Formula Weight 416.3
Percent Purity 96%
Chemical Name or Material Cypermethrin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and
EINECSNumber 257-842-9

RUO – Research Use Only