Recombinant Protein;100UG;Cynomolgus CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag (CDG-C5257) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Gln 23 - Thr 113 (CD3G) (Accession # Q95LI5-1 (CD3E) & Q95LI7-1 (CD3G)).;Cynomolgus CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 40.6 kDa (CD3E) and 40.4 kDa (CD3G). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus and subunit CD3G is fused with a human IgG1 Fc tag at the C-terminus. The reducing (R) heterodimer protein migrates as 45-52 kDa due to glycosylation.;Immobilized Bispecific T cell Engager (CD3 X BCMA) at 1 g/mL (100 L/well) can bind Cynomolgus CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag(Cat. No. CDG-C5257) with a linear range of 0.3-5 ng/mL (QC tested).