missing translation for 'onlineSavingsMsg'
Learn More

Cymoxanil 98.0+%, TCI America™

Catalog No. C26575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26575G 5 g
C265725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26575G Supplier TCI America Supplier No. C26575G

Chemical Identifiers

CAS 57966-95-7
Molecular Formula C7H10N4O3
Molecular Weight (g/mol) 198.182
MDL Number MFCD00137381
InChI Key XERJKGMBORTKEO-VZUCSPMQSA-N
Synonym 1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea, 2-Cyano-N-(ethylcarbamoyl)-2-methoxyiminoacetamide
PubChem CID 5364079
IUPAC Name (1E)-2-(ethylcarbamoylamino)-N-methoxy-2-oxoethanimidoyl cyanide
SMILES CCNC(=O)NC(=O)C(=NOC)C#N

Specifications

Melting Point 162°C
Quantity 5 g
UN Number 3077
Formula Weight 198.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cymoxanil
RTECSNumber AB5957000
TSCA Yes