Learn More
Chemglass Inc Cylinder, Graduated, 250mL, 24/40 Outer Joint
Numéro de catalogue. CG122303
Encompass
Cylinder, Graduated, 250mL, 24/40 Outer Joint. Cylinders are calibrated to contain at 20 C. Supplied without stopper. For standard taper stoppers see CG-3000. Tolerances and subdivisions are the same as CG-8242.
Numéro de catalogue. CG122303 Fournisseur Chemglass Inc Code fournisseur CG122303
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Cylinder, Graduated, 250mL, 24/40 Outer Joint. Cylinders are calibrated to contain at 20 C. Supplied without stopper. For standard taper stoppers see CG-3000. Tolerances and subdivisions are the same as CG-8242.