Learn More
BeLLCo Glass Cylinders are useful for general laboratory procedures and are autoclavable. They have blue graduations to aid in viewing and dispensing of liquids. Material: polypropylene Autoclavable Size: 100ml Dimensions: 34x260mm
Numéro de catalogue. 501945736
Encompass
Cylinders are useful for general laboratory procedures and are autoclavable. They have blue graduations to aid in viewing and dispensing of liquids. Material: polypropylene Autoclavable Size: 100ml Dimensions: 34x260mm
Numéro de catalogue. 501945736 Fournisseur BeLLCo Glass Code fournisseur 562610100
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Cylinders are useful for general laboratory procedures and are autoclavable. They have blue graduations to aid in viewing and dispensing of liquids. Material: polypropylene Autoclavable Size: 100ml Dimensions: 34x260mm