Learn More
Vee Gee Scientific Cylinder, Graduated , Class A, 2000 mL
Numéro de catalogue. NC2386740
Encompass
SIBATA Class A Graduated Cylinder with double-metric scale, graduated, to contain, and calibration temperature of 20C. The graduated cylinder features hexagonal base for added stability* Made out of high-quality borosilicate glass* Double metric scale* Contains durable white enamel graduation markings* Wide range of sizes available (5 mL to 2000 mL)* Feature hexagonal bases for added stability* All sizes (except 2000 mL) come with a protective bumper guard* Calibration Temperature: 68F (20C)* Exceeds ASTM E-1272 Class A Standards * Graduation Interval: 20 mL* Graduation Range: 100.0 to 2000.0 mL* Material: Borosilicate Glass* Scale Type(s): Double Metric* Tolerance: 6.00 mL* Quantity: 1 Each
Numéro de catalogue. NC2386740 Fournisseur Vee Gee Scientific Code fournisseur 2351A2000
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
SIBATA Class A Graduated Cylinder with double-metric scale, graduated, to contain, and calibration temperature of 20C. The graduated cylinder features hexagonal base for added stability* Made out of high-quality borosilicate glass* Double metric scale* Contains durable white enamel graduation markings* Wide range of sizes available (5 mL to 2000 mL)* Feature hexagonal bases for added stability* All sizes (except 2000 mL) come with a protective bumper guard* Calibration Temperature: 68F (20C)* Exceeds ASTM E-1272 Class A Standards * Graduation Interval: 20 mL* Graduation Range: 100.0 to 2000.0 mL* Material: Borosilicate Glass* Scale Type(s): Double Metric* Tolerance: 6.00 mL* Quantity: 1 Each