missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CyDye™ DIGE Fluor Minimal Dye Labeling Kits

Numéro de catalogue. 45002245
Change view
Click to view available options
Quantity:
2 nmol of each: Cy2, Cy3, and Cy5
5 nmol of each: Cy2, Cy3, and Cy5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
45002245 2 nmol of each: Cy2, Cy3, and Cy5
45002256 5 nmol of each: Cy2, Cy3, and Cy5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 45002245 Fournisseur Cytiva Code fournisseur 28934530
Il en reste null

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel. Amersham CyDye™ DIGE Fluor Minimal Labeling Kits contain size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.

  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy

All minimal dyes allow the labeling of 50μg protein per labeling reaction. Minimal Dye Labeling Kits contain 2nmol or 5nmol of each of the minimal dyes.

For general 2-D applications where sufficient amounts of sample are available.

Spécifications

Format Solid
For Use With (Application) The Scarce Sample Labeling Kit is designed to be used for applications in which only small amounts of sample are available, for example, in laser capture microdissection, detects up to 3 prelabeled protein samples, standard on the same 2-D electrophoresis gel
Quantity 2 nmol of each: Cy2, Cy3, and Cy5
Product Type Amersham CyDye DIGE Fluor Minimal Dye Labeling Kit
Kit Contents 2nM of CyDye DIGE Fluor minimal labeling kit (Cy2, Cy3 and Cy5)
Content And Storage -15°C to -30°C
Color Blue, Green, Red