missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmagnesium Bromide (ca. 10% in Tetrahydrofuran, ca. 0.7mol/L), TCI America™

Catalog No. C2039100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2039100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2039100G Supplier TCI America Supplier No. C2039100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23719-80-4
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00672094
InChI Key QUPLLJGAJVXXBW-UHFFFAOYSA-M
Synonym cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide
PubChem CID 11116189
IUPAC Name bromo(cyclopropyl)magnesium
SMILES Br[Mg]C1CC1

Specifications

Quantity 100 g
UN Number 3399
Formula Weight 145.28
Chemical Name or Material Cyclopropylmagnesium Bromide (ca. 10% in Tetrahydrofuran, ca. 0.7mol/L)