missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF, AcroSeal™

Catalog No. AC445958000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC445958000 800 mL AcroSeal™ Glass bottle
AC445951000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445958000 Supplier Thermo Scientific Chemicals Supplier No. 445958000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 23719-80-4 | C3H5BrMg | 145.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23719-80-4 , 96-47-9
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00672094
InChI Key QUPLLJGAJVXXBW-UHFFFAOYSA-M
Synonym cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
PubChem CID 11116189
IUPAC Name magnesium;cyclopropane;bromide
SMILES Br[Mg]C1CC1

Specifications

Flash Point −11°C
Packaging AcroSeal™ Glass bottle
Linear Formula C3H5MgBr
Quantity 800 mL
Solubility Information Solubility in water: reacts
Formula Weight 145.28
Chemical Name or Material Cyclopropylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Causes serious eye irritation.
May cause respiratory irritation.<b
Precautionary Statement GHS P Statement
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye prot

RUO – Research Use Only