missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylamine 95.0+%, TCI America™

Catalog No. C09815ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09815ML 5 mL
C098125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09815ML Supplier TCI America Supplier No. C09815ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 765-30-0
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.096
MDL Number MFCD00001301
InChI Key HTJDQJBWANPRPF-UHFFFAOYSA-N
Synonym cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine
PubChem CID 69828
ChEBI CHEBI:34660
IUPAC Name cyclopropanamine
SMILES C1CC1N

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 50°C
Quantity 5 mL
UN Number 2733
Formula Weight 57.10
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopropylamine
EINECSNumber (3)-4396
RTECSNumber GZ0685500
TSCA Yes