missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropylamine, 98%
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | Cyclopropylamine |
| Point de fusion | -50,0 °C |
| Densité | 0.8240g/mL |
| Point d’ébullition | 49,0 °C à 50,0 °C |
| Point d’éclair | −25 °C |
| Fourchette de pourcentages d’essai | 97,5% min. (GC) |
| Emballage | Bouteille en verre |
| Indice de réfraction | 1,4190 à 1,4220 |
| Formule linéaire | C3H5NH2 |
| Quantité | 25 mL |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Harmful if inhaled. Harmful if swallowed. Causes serious eye damage. Causes severe skin burns and eye damage. Harmful in contact with skin. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 212-142-2 |
RUO – Usage réservé à la recherche