missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl methyl ketone, 99%

Numéro de catalogue. aaa1354014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1354014 25 g
AAA1354022 100 g
AAA1354036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1354014 Fournisseur Thermo Scientific Chemicals Code fournisseur A1354014
Il en reste null

CAS: 765-43-5 | C5H8O | 84.118 g/mol

Cyclopropyl methyl ketone was used in the preparation of homoallylic alcohols. Acetylcyclopropane is used as an intermediate for the manufacturing pharmaceuticals, agrochemicals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl methyl ketone was used in the preparation of homoallylic alcohols. Acetylcyclopropane is used as an intermediate for the manufacturing pharmaceuticals, agrochemicals and other organic compounds.

Solubility
Fully miscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Identifiants chimiques

CAS 765-43-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00001297
InChI Key HVCFCNAITDHQFX-UHFFFAOYSA-N
Synonym cyclopropyl methyl ketone, acetylcyclopropane, methyl cyclopropyl ketone, ethanone, 1-cyclopropyl, 1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, ketone, cyclopropyl methyl, cyclopropylmethylketone, unii-n27yy1xcfh, cyclopropylethanone
PubChem CID 13004
IUPAC Name 1-cyclopropylethanone
SMILES CC(=O)C1CC1

Spécifications

Melting Point <-70°C
Density 0.898
Boiling Point 113°C to 114°C
Flash Point 16°C (61°F)
Refractive Index 1.424
Quantity 25 g
UN Number UN1992
Beilstein 1633817
Solubility Information Fully miscible in water.
Formula Weight 84.12
Percent Purity 99%
Chemical Name or Material Cyclopropyl methyl ketone
Afficher plus de résultats Afficher moins de résultats
Hazard Category H225-H301-H315-H319
Hazard Statement GHS H Statement
H225-H301-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 212-146-4
RTECSNumber KM5648000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only