missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxylic acid chloride, 98%

Catalog No. AC111635000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111635000 500 g Glass bottle
AC111630250 25 g Glass bottle
AC111631000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111635000 Supplier Thermo Scientific Chemicals Supplier No. 111635000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4023-34-1 | C4H5ClO | 104.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4023-34-1
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.54
InChI Key ZOOSILUVXHVRJE-UHFFFAOYSA-N
Synonym cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
PubChem CID 77637
IUPAC Name cyclopropanecarbonyl chloride
SMILES C1CC1C(=O)Cl

Specifications

Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 119.0°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4515 to 1.4535
Linear Formula C3H5COCl
Quantity 500 g
Beilstein 09,4
Specific Gravity 1.15
Solubility Information Solubility in water: decomposes
Formula Weight 104.54
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopropanecarboxylic acid chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
EINECSNumber 223-684-4

RUO – Research Use Only