missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017846
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017846 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017846 Supplier Merck Emd Millipore Supplier No. PHR14041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Cyclophosphamide is one of the cytostatic compounds that is a constituent of several polychemotherapy regimens. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 6055-19-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C7H15 Cl2N2O2 P · H2O
MDL Number MFCD00149395
Melting Point 49°C to 51°C
Synonym Cyclophosphamide monohydrate; 2-[Bis(2-chloroethyl)amino]tetrahydro-2 H-1,3,2-oxazaphosphorine 2-oxide; Cytoxan
Type Certified Reference Material
Quantity 1 g
Formula Weight 279.10
Grade Certified Reference Material
Show More Show Less