missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C24421G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24421G Supplier TCI America Supplier No. C24421G
Only null left

Chemical Identifiers

CAS 63076-51-7
Molecular Formula C5H11BO2
MDL Number MFCD01074541
InChI Key VTTDFSNKIMAQTB-UHFFFAOYSA-N
PubChem CID 2734327
IUPAC Name cyclopentylboronic acid

Specifications

Melting Point 126°C
Quantity 1 g
Synonym cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid
SMILES B(C1CCCC1)(O)O
Molecular Weight (g/mol) 113.951
Formula Weight 113.95
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentylboronic Acid (contains varying amounts of Anhydride)