missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene, 95+%

Catalog No. AC295901000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295901000 100 mL Glass bottle
AC295905000 500 mL Glass bottle
AC295900025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC295901000 Supplier Thermo Scientific Chemicals Supplier No. 295901000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 142-29-0 | C5H8 | 68.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142-29-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.11
MDL Number MFCD00001394
InChI Key LPIQUOYDBNQMRZ-UHFFFAOYSA-N
Synonym 1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
PubChem CID 8882
ChEBI CHEBI:49155
IUPAC Name cyclopentene
SMILES C1CC=CC1

Specifications

Melting Point -135.0°C
Color Colorless
Density 0.7740g/mL
Boiling Point 44.0°C
Flash Point −30°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4200 to 1.4240
Linear Formula C5H8
Quantity 100 mL
Beilstein 05, 61
Specific Gravity 0.774
Solubility Information Solubility in water: immiscible
Viscosity 0.36 mPa.s (15°C)
Formula Weight 68.11
Percent Purity 95+%
Grade Technical
Physical Form Liquid
Chemical Name or Material Cyclopentene, tech., ≥95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediate
EINECSNumber 205-532-9
RTECSNumber GY5950000
TSCA TSCA

RUO – Research Use Only