missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene-1-boronic acid, 97%

Catalog No. AAH52531MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52531MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH52531MD Supplier Thermo Scientific Chemicals Supplier No. H52531MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 850036-28-1 | C5H9BO2 | 111.935 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 850036-28-1
Molecular Formula C5H9BO2
Molecular Weight (g/mol) 111.935
MDL Number MFCD02179496
InChI Key UZBHNSVUMGIKLU-UHFFFAOYSA-N
Synonym cyclopent-1-en-1-ylboronic acid, 1-cyclopentenylboronic acid, cyclopentenylboronic acid, cyclopent-1-en-1-yl boronic acid, cyclopentene-1-boronic acid, cyclopenten-1-yl boronic acid, cyclopent-1-ene-1-boronic acid, boronic acid, 1-cyclopenten-1-yl, cyclopenteneboronic acid, acmc-209pzb
PubChem CID 3861343
IUPAC Name cyclopenten-1-ylboronic acid
SMILES B(C1=CCCC1)(O)O

Specifications

Melting Point 134°C to 140°C
Quantity 250 mg
Formula Weight 111.94
Percent Purity 97%
Chemical Name or Material Cyclopentene-1-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only