missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone Oxime 98.0+%, TCI America™

Catalog No. C130725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C130725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C130725G Supplier TCI America Supplier No. C130725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1192-28-5
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00001420
InChI Key YGNXYFLJZILPEK-UHFFFAOYSA-N
Synonym cyclopentanone oxime, cyclopentanoneoxime, cyclopentanone, oxime, unii-qy3bv00pcy, ccris 7163, qy3bv00pcy, n-1e-cyclopentylidene hydroxylamine, hydroxyimino cyclopentane, cyclopentanone,oxime, hydroxyiminocyclopentane
PubChem CID 14500
IUPAC Name N-cyclopentylidenehydroxylamine
SMILES ON=C1CCCC1

Specifications

Melting Point 58°C
Color White
Boiling Point 97°C
Quantity 25 g
Formula Weight 99.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentanone Oxime
RTECSNumber GY5150000
TSCA Yes