missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienyltitanium(IV) Trichloride 98.0+%, TCI America™

Catalog No. C19945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19945G 5 g
C19941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19945G Supplier TCI America Supplier No. C19945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1270-98-0
Molecular Formula C5H5Cl3Ti
Molecular Weight (g/mol) 219.31
InChI Key QOXHZZQZTIGPEV-UHFFFAOYSA-K
Synonym Trichloro(cyclopentadienyl)titanium(IV)

Specifications

Melting Point 210°C
Physical Form Crystalline Powder
Quantity 5 g
UN Number 3261
Formula Weight 219.31
Percent Purity ≥98.0% (T)
Grade Reagent
Chemical Name or Material Cyclopentadienyltitanium(IV) Trichloride
RTECSNumber XR1927300
TSCA No