missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienylbis(triphenylphosphine)ruthenium(II) Chloride, TCI America™

Catalog No. C22011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22011G Supplier TCI America Supplier No. C22011G
Only null left

Chemical Identifiers

CAS 32993-05-8
Molecular Formula C43H41ClOP2Ru
Molecular Weight (g/mol) 772.27
MDL Number MFCD00075004
InChI Key YKZUFRITHQLOHK-UHFFFAOYSA-M
Synonym bis triphenylphosphine chloro cyclopentadienylruthenium, chlorocyclopentadienylbis triphenylphosphine ruthenium ii 095h
PubChem CID 132285030
IUPAC Name Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
SMILES Cl[Ru++].CCO.c1cccc1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C
Quantity 1 g
Formula Weight 726.20
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentadienylbis(triphenylphosphine)ruthenium(II) Chloride