missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclooctanone 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 45°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 126.20 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Cyclooctanone |
Safety and Handling
| RTECSNumber | GX9800000 |
| TSCA | Yes |