missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctane, 99+%

Catalog No. AAL15000AK
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15000AK 250 mL
AAL15000AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15000AK Supplier Thermo Scientific Chemicals Supplier No. L15000AK
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 292-64-8 | C8H16 | 112.216 g/mol

Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 292-64-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00004162
InChI Key WJTCGQSWYFHTAC-UHFFFAOYSA-N
Synonym octamethylene, cyclooctan, unii-kkz3kbs654, kkz3kbs654, cyclooctane, acmc-209h7x, cambridge id 6337558, cambridge id 6511221, cambridge id 6544558, cyclooctane, analytical standard
PubChem CID 9266
IUPAC Name cyclooctane
SMILES C1CCCCCCC1

Specifications

Melting Point 12°C to 14°C
Density 0.835
Boiling Point 150°C to 152°C
Flash Point 28°C (82°F)
Refractive Index 1.458
Quantity 250 mL
UN Number UN3295
Beilstein 1900349
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 112.22
Percent Purity ≥99%
Chemical Name or Material Cyclooctane
Show More Show Less
Hazard Category H226-H304
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 206-031-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only