missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyltrichlorosilane 98.0+%, TCI America™

Catalog No. p-7124668
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08925G 5 g
C089225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08925G Supplier TCI America Supplier No. C08925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-12-4
Molecular Formula C6H11Cl3Si
Molecular Weight (g/mol) 217.589
MDL Number MFCD00021280
InChI Key SIPHWXREAZVVNS-UHFFFAOYSA-N
Synonym cyclohexyltrichlorosilane, trichloro cyclohexyl silane, silane, trichlorocyclohexyl, cyclohexane, trichlorosilyl, cyclohexane, 1-trichlorosilyl, cyclohexyl trichlorosilane, trichlorocyclohexyl silane, silane, cyclohexyltrichloro, 1-trichlorosilyl cyclohexane, trichlorocyclohexylsilane
PubChem CID 60987
IUPAC Name trichloro(cyclohexyl)silane
SMILES C1CCC(CC1)[Si](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 207°C
Quantity 5 g
UN Number 1763
Formula Weight 217.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyltrichlorosilane