missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine hydrobromide, 98%

Catalog No. AAA1843936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1843936 500 g
AAA1843922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1843936 Supplier Thermo Scientific Chemicals Supplier No. A1843936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26227-54-3 | C6H14BrN | 180.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26227-54-3
Molecular Formula C6H14BrN
Molecular Weight (g/mol) 180.089
MDL Number MFCD00034986
InChI Key QOHWJRRXQPGIQW-UHFFFAOYSA-N
Synonym cyclohexylamine hydrobromide, cyclohexanamine hydrobromide, cyclohexylammonium bromide, cyclohexanamine, hydrobromide, cyclohexanaminium bromide, cyclohexyl amine hydrobromide, acmc-1ccru, cyclohexylamine hbr, ksc268i9n, cyclohexamine, hydrobromide salt
PubChem CID 117303
IUPAC Name cyclohexanamine;hydrobromide
SMILES C1CCC(CC1)N.Br

Specifications

Melting Point 197°C to 201°C
Quantity 500 g
Beilstein 3908098
Sensitivity Hygroscopic
Formula Weight 180.09
Percent Purity 98%
Chemical Name or Material Cyclohexylamine hydrobromide
Hazard Category H302-H315-H319-H335-H361
Hazard Statement GHS H Statement
H301-H361-H315-H319-H335
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 247-526-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only