missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methyl sulfide, 97%

Catalog No. AAL0371206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0371206 5 g
AAL0371214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0371206 Supplier Thermo Scientific Chemicals Supplier No. L0371206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7133-37-1 | C7H14S | 130.25 g/mol

Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

Solubility
Soluble in water 173.5 mg/L @ 25°C, alcohol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 7133-37-1
Molecular Formula C7H14S
Molecular Weight (g/mol) 130.25
MDL Number MFCD00039460
InChI Key QQBIOCGHCKNYGP-UHFFFAOYSA-N
Synonym cyclohexyl methyl sulfide, methylthio cyclohexane, sulfide, cyclohexyl methyl, cyclohexane, methylthio, methylthiocyclohexane, methylcyclohexylsulfide, cyclohexylmethylsulphide, cyclohexyl methylsulfide, methyl cyclohexyl sulfide, methylsulfanyl cyclohexane
PubChem CID 81553
SMILES CSC1CCCCC1

Specifications

Melting Point -80°C
Density 0.944
Boiling Point 89°C to 90°C (35 mmHg)
Flash Point 61°C (142°F)
Odor Unpleasant
Refractive Index 1.494
Quantity 5 g
UN Number UN3334
Beilstein 1919309
Solubility Information Soluble in water 173.5mg/L @ 25°C,alcohol.
IUPAC Name (methylsulfanyl)cyclohexane
Formula Weight 130.25
Percent Purity 97%
Chemical Name or Material Cyclohexyl methyl sulfide
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber 230-431-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only