missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexyl(dimethoxy)methylsilane 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 196°C |
| Quantity | 500 mL |
| UN Number | 3082 |
| Formula Weight | 188.34 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Cyclohexyl(dimethoxy)methylsilane |
Safety and Handling
| EINECSNumber | (3)-4027 |
| TSCA | Yes |