missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog No. AAL1292514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1292514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1292514 Supplier Thermo Scientific Chemicals Supplier No. L1292514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3066-71-5 | Molecular Formula: C9H14O2 | Molecular Weight: 154.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3066-71-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.21
MDL Number MFCD00046349
InChI Key KBLWLMPSVYBVDK-UHFFFAOYSA-N
Synonym cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference
PubChem CID 18298
IUPAC Name cyclohexyl prop-2-enoate
SMILES C=CC(=O)OC1CCCCC1

Specifications

Density 0.98
Boiling Point 182°C to 184°C
Flash Point 68°C (154°F)
Refractive Index 1.459
Quantity 25 g
UN Number UN3082
Beilstein 1937528
Formula Weight 154.21
Percent Purity ≥98%
Chemical Name or Material Cyclohexyl acrylate, stabilized with 100ppm 4-methoxyphenol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H335-H227
Causes skin irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 221-319-3
RTECSNumber AS7350000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only