missing translation for 'onlineSavingsMsg'
Learn More

Acétate de cyclohexyle 99,0+%, TCI America™

Catalog No. A0028500ML
Change view
Click to view available options
Quantité:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
A0028500ML 500 mL
A002825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0028500ML Supplier TCI America Supplier No. A0028500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 622-45-7
Formule moléculaire C8H14O2
Poids moléculaire (g/mol) 142.198
Numéro MDL MFCD00003850
Clé InChI YYLLIJHXUHJATK-UHFFFAOYSA-N
Synonyme acetic acid, cyclohexyl ester, hexalin acetate, cyclohexanyl acetate, cyclohexanol, acetate, cyclohexanolazetat, adronal acetate, acetic acid cyclohexyl ester, cyclohexane acetate, cyclohexylester kyseliny octove, unii-ul0rs4h1ue
PubChem CID 12146
ChEBI CHEBI:31447
Nom de l’IUPAC Acétate de cyclohexyle
SOURIRES CC(=O)OC1CCCCC1

Specifications

Nom ou substance chimique Cyclohexyl Acetate
Couleur Incolore
Point d’ébullition 174 °C
Quantité 500 mL
Numéro UN 2243
Poids de formule 142.20
Pourcentage de pureté ≥99,0% (GC)
Forme physique Liquide
EINECSNumber (3)-2316&(3)-2359
RTECSNumber AG5075000
TSCA Oui