missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Cycloheximide, InSolution™, Calbiochem™,

Catalog No. 2397651ML
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Alu drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2397651ML 1 mL Alu drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2397651ML Supplier MilliporeSigma™ Supplier No. 2397651ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recommended Storage

2°C to 8°C

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

Packaging Alu drum
Quantity 1 mL
Physical Form Liquid
Recommended Storage 2°C to 8°C