missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene, 98%, Spectrum™ Chemical

Catalog No. 18610114
Change view
Click to view available options
Quantity:
1 L
200 L
Packaging:
Amber Glass Bottle
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610114 200 L Drum
18610113 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610114 Supplier Spectrum Chemical Mfg Cor Supplier No. C1464200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-83-8
Molecular Weight (g/mol) 82.15

Specifications

CAS Max % 1
Quantity 200 L
Packaging Drum
Boiling Point 83°C
Percent Purity 0.98
Grade Ungraded
Chemical Name or Material Cyclohexene

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.