missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone 99.0+%, TCI America™

Numéro de catalogue. p-7124462
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C048925ML 25 mL
C0489500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C048925ML Fournisseur TCI America Code fournisseur C048925ML
Il en reste null

Identifiants chimiques

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001625
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1

Spécifications

Color Yellow
Boiling Point 157°C
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1915
Formula Weight 98.15
Percent Purity ≥99.0% (GC)
Chemical Name or Material Cyclohexanone
EINECSNumber (3)-2376
RTECSNumber GW1050000
TSCA Yes