missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, ACS, 99%, Spectrum™ Chemical

Numéro de catalogue. p-7165379
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18610105 100 mL Amber Glass Bottle
18610104 500 mL Amber Glass Bottle
18610106 1 L Amber Glass Bottle
18610107 4 L Amber Glass Bottle
18610102 20 L Steel Can
18610103 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. 18610105 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur C1462100ML
Il en reste null

Identifiants chimiques

CAS 108-94-1

Spécifications

CAS Max % 1
Boiling Point 156°C
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.0005
Water 0.0005
Percent Purity 0.99
Grade Reagent
Chemical Name or Material Cyclohexanone

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.