missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, 99+%, ACS reagent

Catalog No. AC406092500
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC406092500 250 mL
AC406090010 1 L
AC406090025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC406092500 Supplier Thermo Scientific Chemicals Supplier No. 406092500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-94-1 | C6H10O | 98.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1

Specifications

Melting Point -47.0°C
Density 0.9470g/mL
Quantity 250 mL
Flash Point 43°C
Refractive Index 1.4450 to 1.4560
Specific Gravity 0.947
Linear Formula C6H10(=O)
Water 0.05% max.
Boiling Point 155.0°C
Solubility Information Solubility in water: 150g/L (10°C). Other solubilities: soluble in all common organic solvents
Formula Weight 98.14
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Cyclohexanone
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye damage. Causes skin irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only