missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanol, 97%, Spectrum™ Chemical

Catalog No. 18610100
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610100 4 L Steel Can
18610098 20 L Steel Can
18610099 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610100 Supplier Spectrum Chemical Mfg Cor Supplier No. C14604LTGL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-82-7
Molecular Weight (g/mol) 100.16

Specifications

CAS Max % 1
Quantity 4 L
Packaging Steel Can
Boiling Point 161.84°C
Percent Purity 97%
Grade Reagent
Chemical Name or Material Cyclohexanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.