missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanethiol 98.0+%, TCI America™

Numéro de catalogue. C072725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C072725ML 25 mL
C0727500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C072725ML Fournisseur TCI America Code fournisseur C072725ML
Il en reste null

Identifiants chimiques

CAS 1569-69-3
Molecular Formula C6H12S
Molecular Weight (g/mol) 116.222
MDL Number MFCD00001455
InChI Key CMKBCTPCXZNQKX-UHFFFAOYSA-N
Synonym cyclohexyl mercaptan, cyclohexyl thiol, cyclohexylmercaptan, cyclohexylthiol, cyklohexylmerkaptan, cyklohexanthiol, cyklohexanthiol czech, cyklohexylmerkaptan czech, hexahydrothiophenol, mercaptocyclohexane
PubChem CID 15290
IUPAC Name cyclohexanethiol
SMILES C1CCC(CC1)S

Spécifications

Color Colorless
Boiling Point 158°C
Quantity 25 mL
UN Number 3054
Formula Weight 116.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclohexanethiol
EINECSNumber (3)-2303
RTECSNumber GV7525000
TSCA Yes