missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanemethanol, 99%

Catalog No. AAL0653814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0653814 25 g
AAL0653822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0653814 Supplier Thermo Scientific Chemicals Supplier No. L0653814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-49-2 | C7H14O | 114.188 g/mol

Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.

Solubility
Soluble in Ether, Alcohols.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Acid chlorides, Acid anhydrides.

Chemical Identifiers

CAS 100-49-2
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00001510
InChI Key VSSAZBXXNIABDN-UHFFFAOYSA-N
Synonym cyclohexanemethanol, cyclohexylcarbinol, hexahydrobenzyl alcohol, cyclohexanecarbinol, methanol, cyclohexyl, cyclohexylmethyl alcohol, benzyl alcohol, hexahydro, usaf do-49, hydroxymethylcyclohexane, hydroxymethyl cyclohexane
PubChem CID 7507
IUPAC Name cyclohexylmethanol
SMILES C1CCC(CC1)CO

Specifications

Density 0.926
Boiling Point 180°C to 181°C
Flash Point 71°C (159°F)
Refractive Index 1.464
Quantity 25 g
Beilstein 773712
Merck Index 14,2731
Solubility Information Soluble in Ether,Alcohols.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material Cyclohexanemethanol
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 202-857-8
RTECSNumber GV5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only