missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane 99.5+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 100 mL |
| Physical Form | Clear Liquid at 20°C |
| UN Number | 1145 |
| Boiling Point | 81°C |
| Formula Weight | 84.16 |
| Percent Purity | ≥99.5% (GC) |
| Chemical Name or Material | Cyclohexane [for Spectrophotometry] |
Safety and Handling
| EINECSNumber | (3)-2233 |
| RTECSNumber | GU6300000 |
| TSCA | Yes |