missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane 99.5+%, TCI America™

Catalog No. C0818500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0818500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0818500ML Supplier TCI America Supplier No. C0818500ML
Only null left

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Color Colorless
Quantity 500 mL
Physical Form Clear Liquid at 20°C
UN Number 1145
Boiling Point 81°C
Formula Weight 84.16
Percent Purity ≥99.5% (GC)
Chemical Name or Material Cyclohexane [for HPLC Solvent]
RTECSNumber GU6300000
TSCA Yes