missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, MilliporeSigma™

Catalog No. MCX22861
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MCX22861 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MCX22861 Supplier Technidata Supplier No. CX22861

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For environmental analysis

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.7°C
Color Colorless
Density 0.78g/cm3 at 20°C
Quantity 4 L
Explosion Limit 1.2 to 8.3%(V)
Flash Point -20.15°C (-4.3°F)
Odor Pungent, fruity
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH2(CH2)4CH2
Vapor Pressure 103 hPa (20°C)
UN Number UN 1145
Boiling Point 80.6°C
Solubility Information Insoluble in water
Formula Weight 84.18
Percent Purity ≥99.0%
Chemical Name or Material Cyclohexane
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Store at room temperature.