missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7%, Solstice
Description
- Solstice has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | 4°C to 7°C (literature) |
| Absorbance | max. 0.10 at 235 nm, max. 0.20 at 230 nm, max. 0.50 at 220 nm, max. 1.00 at 210 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.01 at 255 nm, max. 0.03 at 250 nm, max. 0.08 at 240 nm |
| CAS Max % | ≤100.0000% |
| Density | 0.779 g/mL at 25 °C(lit.); 0.780 g/mL at 25 °C |
| Quantity | 4 x 2.5 L |
| Flash Point | −18°C |
| Packaging | Glass Bottle |
| Linear Formula | C6H12 |
| UN Number | UN1145 |
| Beilstein | 1900225 |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Cyclohexane |
| EINECSNumber | 203-806-2 |
| Recommended Storage | Room Temp |