missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Supplier:  Honeywell-Burdick and Jackson 0534

 View more versions of this product

Catalog No. 6000482

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cyclohexane
110-82-7
100.0000%
80.72°C
−20°C
C6H12
UN1145
1900225
0.6% in Water at 20°C
C1CCCCC1
84.16
CHEBI:29005
>99.5%
B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.5%
0.17 AU max. at 225 nm (UV), 0.02 AU max. at 250 nm (UV), 0.005 AU max. at 300 nm (UV), 0.005 AU max. at 400 nm (UV)
0.78 g/cm3
4 x 4 L
Glass Bottle
MFCD00003814
0.01% max.
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.16
HPLC

Chemical Identifiers

110-82-7
84.16
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
C6H12
MFCD00003814
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Cyclohexane

SDS
Documents

Documents

Promotions

Promotions