missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, ≥99.8%, Laboratory Reagent, Honeywell™
Laboratory Reagent,≥99.8%
Supplier: Honeywell Chemicals C10030725L
Description
- ≥95% for general laboratory use.
Specifications
| Cyclohexane | |
| 110-82-7 | |
| 0.779g/mL at 25°C | |
| 2.5 L | |
| 500°F | |
| Glass Bottle | |
| C6H12 | |
| 168.8mmHg (37.7°C), 77mmHg (20°C) | |
| 1900225 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| cyclohexane | |
| 8078 | |
| 84.16g/mol |
| 6.4°C | |
| Colorless | |
| 81°C (1013 hPa) | |
| 0.09 | |
| Characteristic | |
| Liquid | |
| MFCD00003814 | |
| UN1145 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| C1CCCCC1 | |
| 84.16 | |
| CHEBI:29005 | |
| ≥99.8% |
Chemical Identifiers
| 110-82-7 | |
| 84.16 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| 8078 | |
| cyclohexane |
| C6H12 | |
| MFCD00003814 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| CHEBI:29005 | |
| C1CCCCC1 |
Safety and Handling
P210-P261-P273-P301 + P310-P331-P501
DOTInformation : class 3 - PG 2 - Cyclohexane
RTECSNumber : 203-806-2