missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99.0% min., ACS, MilliporeSigma™

Catalog No. MCX229030
Change view
Click to view available options
Quantity:
1 L
4 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX229030 200 L
MCX22901 1 L
MCX22903 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MCX229030 Supplier Technidata Supplier No. CX229030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Quantity 200 L
Physical Form Liquid
Assay Percent Range 99.0%, min.
Linear Formula -CH2(CH2)4CH2-
Formula Weight 84.16
Grade ACS/NF/USP
Chemical Name or Material Cyclohexane, For ACS analysis