missing translation for 'onlineSavingsMsg'
Learn More

β-Cyclodextrin Pharmaceutical Secondary Standard, 99.9%, MilliporeSigma™ Supelco™

Catalog No. 111015441
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015441 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015441 Supplier Merck Emd Millipore Supplier No. PHR14391G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Beta-Cyclodextrin is the most abundant and cheap cyclic oligosaccharide that forms inclusion complexes with several drug molecules. Its main application is in tablet and capsule formulations.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 7585-39-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Powder
Linear Formula C42H70O35
MDL Number MFCD00078139
Melting Point 290°C to 300°C
Synonym beta-Cyclodextrin; Caraway; Cycloheptaamylose; Cyclomaltoheptaose; Schardinger beta-Dextrin
Type Certified Reference Material
Quantity 1 g
Formula Weight 1134.98
Percent Purity 99.9%
Grade Certified Reference Material
Show More Show Less