missing translation for 'onlineSavingsMsg'
Learn More

gamma-Cyclodextrin hydrate, 99%

Catalog No. AC229905000
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229905000 500 mg Glass bottle
AC229900010 1 g Glass bottle
AC229900050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC229905000 Supplier Thermo Scientific Chemicals Supplier No. 229905000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 91464-90-3
Molecular Formula C48H80O40
Molecular Weight (g/mol) 1297.13
MDL Number MFCD00149574
InChI Key GDSRMADSINPKSL-UHFFFAOYNA-N
Synonym gamma-cyclodextrin hydrate, 1297.12 anhydrous, gamma-cyclodextrin hydrate, vetec tm reagent grade
PubChem CID 71311509
IUPAC Name 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Melting Point 267.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 500 mg
Merck Index 14, 2718
Specific Rotation + 177.00
Solubility Information Solubility in water: 22.3 g/100 mL (25°C).
Formula Weight 1297.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material γ-Cyclodextrin hydrate
Show More Show Less

RUO – Research Use Only