missing translation for 'onlineSavingsMsg'
Learn More

β-Cyclocitral, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014197
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014197 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014197 Supplier Merck Emd Millipore Supplier No. 169761ML
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 432-25-7
Density 0.943 g/mL (at 25°C)
Linear Formula C10H16 O
MDL Number MFCD00079078
Boiling Point 62°C to 63°C (3 mmHg)
Synonym 2,6,6-Trimethyl-1-cyclohexene-1-carboxaldehyde
Type Analytical Standard
Quantity 1 mL
Formula Weight 152.23
Percent Purity ≥97% (GC)
Grade Analytical Standard
Show More Show Less