missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyclobenzaprine hydrochloride

Catalog No. AC466250050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466250050 5 g
AC466250010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466250050 Supplier Thermo Scientific Chemicals Supplier No. 466250050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6202-23-9 | C20H21N·HCl, C20H22ClN | 311.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6202-23-9
Molecular Formula C20H21N·HCl

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: soluble. Other solubilities: soluble in methanol, ethanol
Melting Point 216.0°C to 218.0°C
Formula Weight 311.85
Chemical Name or Material Cyclobenzaprine hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

EINECSNumber 228-264-4

RUO – Research Use Only