accessibility menu, dialog, popup

UserName

CAS RN 6202-23-9

IUPAC Name: hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}propyl)amine chloride
Synonyms: cyclobenzaprine
Molecular Weight (g/mol): 311.85
Molecular Formula: C20H22ClN
InChi Key: VXEAYBOGHINOKW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2C=CC2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Grade 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)