missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorure cyanurique 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 147°C |
| Color | Blanc |
| Quantity | 25 g |
| UN Number | 2670 |
| Formula Weight | 184.4 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Cyanuric Chloride |
Sécurité et manipulation
| EINECSNumber | (5)-1045 |
| RTECSNumber | XZ1400000 |
| TSCA | Yes |