missing translation for 'onlineSavingsMsg'
Learn More

Cyanoacetic Acid 98.0+%, TCI America™

Numéro de catalogue. p-7124420
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C043925G 25 g
C0439500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C043925G Fournisseur TCI America Code fournisseur C043925G
Il en reste null

Identifiants chimiques

CAS 372-09-8
Molecular Formula C3H3NO2
Molecular Weight (g/mol) 85.062
MDL Number MFCD00002677
InChI Key MLIREBYILWEBDM-UHFFFAOYSA-N
Synonym cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova
PubChem CID 9740
ChEBI CHEBI:51889
IUPAC Name 2-cyanoacetic acid
SMILES C(C#N)C(=O)O

Spécifications

Melting Point 71°C
Color White
Boiling Point 108°C
Quantity 25 g
UN Number 3261
Formula Weight 85.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyanoacetic Acid
EINECSNumber (2)-1548
RTECSNumber AG3675000
TSCA Yes